96 Well Plate, RCH PSAXS
Leverancier: Tecan
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Cerex®
TCAT606-0501RCHEA 0 EUR
TCAT606-0501RCH
96 Well Plate, RCH PSAXS
Microplaten Platen voor vastefase-extractie
Mixed mode reverse-phase with an anion exchanger for acidic compounds.
  • Ion-exchange
  • Liquid handler compatible
A 10 µm silica micro particulate with strong anionic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample prep.