Cerex® TRACE-N SPE Columns
Leverancier: Tecan
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
610-0101R 610-0201R 610-315C
TCAT610-0101REA 0 EUR
TCAT610-0101R TCAT610-0201R TCAT610-315C
Cerex® TRACE-N SPE Columns
Kolommen voor chromatografie
End-capped C18 chemistry.
  • Reversed-phase
  • Liquid handler compatible
A 10 µm micro particulate with end-capped C18 chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample prep.