Atlas® NBE SCX SPE Columns
Leverancier: Tecan
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
30174726 30174734
TCAT30174726EA 0 EUR
TCAT30174726 TCAT30174734
Atlas® NBE SCX SPE Columns
Kolommen voor chromatografie
A hydrophilic/lipophilic balanced strong cationic chemistry offering excellent selectivity to basic and neutral compounds. Also utilising Tecan's unique NBE column platform. A direct alternative to Waters Oasis® MCX and Phenomenex Strata XC.
  • Mode: Ion-exchange
  • Liquid handler compatible
Atlas® is a hydrophilic/lipophilic balanced polymeric sorbent with a N-vinylpyrrolidone-divinyl-ben-zene copolymer. Atlas® SCX which incorporates the Atlas’s copolymer technology along with a strong cationic exchange chemistry to provide a focused clean up when working with basic compounds in tough matrices.