SPE Columns, NBE HPSCX
Leverancier: Tecan
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
427-0021R-NBE 427-0051R-NBE
TCAT427-0021R-NBEEA 0 EUR
TCAT427-0021R-NBE TCAT427-0051R-NBE
SPE Columns, NBE HPSCX
Kolommen voor chromatografie
Mixed mode sorbent with both reverse-phase and strong cationic exchange chemistries.
  • Ion-exchange
  • Liquid handler compatible
Its high surface area provides excellent capacity and a high selectivity for basic analytes. Ideal for drug monitoring, screening, identification, confirmation and quantitation.