CRS SPure™ H₂O Gas Filters
Leverancier: Trajan Scientific and Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
202298 202238-B 202238D-B 202238D-SS 202238L-B 202238L-SS 202238-SS 202238XL-B 202238XL-SS 202239-B 202239D-B 202239D-SS 202239L-B 202239L-SS 202239-SS 202239XL-B 202239XL-SS 202243 202245 202247 202238-4-PC 202238-6-PC 202238-B-QC 202238D-4-PC 202238D-6-PC 202238D-B-QC 202238DRP-B-QC 202238DRP-SS-Q 202238D-SS-QC 202238L-4-PC 202238L-6-PC 202238L-B-QC 202238LRP-B-QC 202238LRP-SS-Q 202238L-SS-QC 202238RP-B-QC 202238RP-SS-QC 202238-SS-QC 202238XL-4-PC 202238XL-6-PC 202238XL-B-QC 202238XLRP-B-Q 202238XLRP-SS- 202238XL-SS-QC 202239-B-QC 202239D-B-QC 202239D-SS-QC 202239L-B-QC 202239L-SS-QC 202239-SS-QC 202239XL-B-QC 202239XL-SS-QC
SGEA202298EA 89.8 EUR
SGEA202298 SGEA202238-B SGEA202238D-B SGEA202238D-SS SGEA202238L-B SGEA202238L-SS SGEA202238-SS SGEA202238XL-B SGEA202238XL-SS SGEA202239-B SGEA202239D-B SGEA202239D-SS SGEA202239L-B SGEA202239L-SS SGEA202239-SS SGEA202239XL-B SGEA202239XL-SS SGEA202243 SGEA202245 SGEA202247 SGEA202238-4-PC SGEA202238-6-PC SGEA202238-B-QC SGEA202238D-4-PC SGEA202238D-6-PC SGEA202238D-B-QC SGEA202238DRP-B-QC SGEA202238DRP-SS-Q SGEA202238D-SS-QC SGEA202238L-4-PC SGEA202238L-6-PC SGEA202238L-B-QC SGEA202238LRP-B-QC SGEA202238LRP-SS-Q SGEA202238L-SS-QC SGEA202238RP-B-QC SGEA202238RP-SS-QC SGEA202238-SS-QC SGEA202238XL-4-PC SGEA202238XL-6-PC SGEA202238XL-B-QC SGEA202238XLRP-B-Q SGEA202238XLRP-SS- SGEA202238XL-SS-QC SGEA202239-B-QC SGEA202239D-B-QC SGEA202239D-SS-QC SGEA202239L-B-QC SGEA202239L-SS-QC SGEA202239-SS-QC SGEA202239XL-B-QC SGEA202239XL-SS-QC
CRS SPure™ H₂O Gas Filters
Filters Gasfilters
Pure gas is a crictical requirement in gas chromatography, spectroscopy, optics, lithography and numerous other applications in manufacturing and laboratories. The SPure™ H₂O filters removes moisture, oil and dust from air and inert gases.
  • High-strength polycarbonate and acrylic bodies with aluminum end caps
  • Available with standard brass and stainless steel compression fittings
  • It is suitable for general purpose, non-critical laboratory use
  • Maximum pressure 6,89 bar
It is not recommended for house compressed air lines or for GC carrier gases and not use for with hazardous, flammable, or reactive gases.