Crimpers and Decappers
Leverancier: Trajan Scientific and Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
308990 311990 311991 313990 313991 313992 320990 320991 320992
SGEA308990EA 231 EUR
SGEA308990 SGEA311990 SGEA311991 SGEA313990 SGEA313991 SGEA313992 SGEA320990 SGEA320991 SGEA320992
Crimpers and Decappers
Capsuleer- en decapsuleertangen
Crimping tools have advanced the standards of manual crimpers and decappers by addressing all of the deficiencies associated with traditional blocky, metal tools.
  • Reliabily seal crimps caps manually with less physical strain
  • constructed of tough, light weight plastic that is incredibly durable but weighs much less than metal crimping tools
  • Curved handles create a natural grip, greatly improving comfort over metal grips
  • Unique bottom pull technology allows for steadier operation in comparison to top-push handle designs
Application: Low volume crimping/decapping applications for aluminium crimp caps.
The adjustment knob is easily viewed and clearly displays + and − symbols along with directional arrows, simplifying the process of adjusting the crimp setting - no tools needed.
No more extra squeeze required. When the adjustment knob touches the body of the crimper, the desired crimp settling has been reached - significantly reducing arm strain and fatigue associated with traditional manual crimping.