Atlas® RCH SCX SPE Plates, 96-well
Leverancier: Tecan
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
30174700 30174708
TCAT30174700EA 0 EUR
TCAT30174700 TCAT30174708
Atlas® RCH SCX SPE Plates, 96-well
Microplaten Platen voor vastefase-extractie
A hydrophilic/lipophilic balanced strong cationic chemistry offering excellent selectivity to basic and neutral compounds. A direct alternative to waters Oasis® MCX and Phenomenex Strata XC.
  • Base resin chemistry: N-vinylpyrrolidonedivinylbenzene copolymer
  • Surface chemistry: sulfonic acid
  • Particle packing: monodisperse porous microsphere
  • pH stability: 1 - 14
Tecan’s new addition to the Resolvex Atlas® smart consumables family is a strong cation exchange chemistry using a balanced hydrophilic-lipophilic base resin. Resolvex Atlas® is a hydrophilic-lipophilic balanced polymeric sorbent with a N-vinylpyrrolidone-divinylbenzene copolymer. this incorporates this copolymer
technology along with a strong cationic exchange hemistry to provide a focused clean-up when working with basic compounds in tough matrices. In addition to the balanced copolymer technology it provides an additional chemistry that allows for strong retention of basic compounds.
Ideal for use in small molecules and pharmaceutical drugs, sample preparation for drug monitoring, screening, peptide clean-up, etc., biopharma/life science and food safety.