SPE Plates, 96-well, NBE OFXQ
Leverancier: Tecan
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
635-0031NBE 635-0051NBE
TCAT635-0031NBEEA 0 EUR
TCAT635-0031NBE TCAT635-0051NBE
SPE Plates, 96-well, NBE OFXQ
Microplaten Platen voor vastefase-extractie
Proprietary blend of ion-exchange and water-wettable polymer targeted towards oral fluid collection devices.
  • Ion-exchange
  • Liquid handler compatible
A proprietary polymer resin blend of ion exchange chemistry providing unique selectivity. Especially designed for scientists working with oral fluid matrixes.