SPE Plates, 96-well, NBE WWP2
Leverancier: Tecan
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Cerex®
TCAT417-0051NBEEA 0 EUR
TCAT417-0051NBE
SPE Plates, 96-well, NBE WWP2
Microplaten Platen voor vastefase-extractie
Polymeric water-wettable resin ideal for reverse-phase retention of aqueous samples.
  • Reversed-phase
  • Liquid handler compatible
Special water wettable polymer with 300 Å pore size ideal for protein digest clean up and other protein related workflows.