Scotch-Brite™ Diamond Floor Brushes
Leverancier: 3M
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
DIA1000432 DIA3000432
MMMADIA1000432EA 1470 EUR
MMMADIA1000432 MMMADIA3000432
Scotch-Brite™ Diamond Floor Brushes
Borstels
Long-lasting, conformable diamond bristle brush designed to clean and polish uncoated concrete in one step. For daily use on low speed floor equipment.
  • Easily and quickly replaceable
  • No pre-soaking required
  • Bladeless installation
  • No assembly required
Two grit levels of the Scotch-Brite™ Diamond Floor Brush are available:
• Scotch-Brite™ Diamond Floor Brush 1000
• Scotch-Brite™ Diamond Floor Brush 3000
This product is designed to be used as part of the 3M™ Concrete Floor Polishing and Maintenance System alongside the 3M™ Concrete Cleaner and Densifier.
Designed to clean and polish uncoated concrete floors. Can improve concrete floors’ gloss and clarity/distinctness of image (DOI). Flexible bristles conform to uneven floors providing uniform appearance and cleanliness. Significantly longer life and durability compared with traditional and competitive diamond floor pads. Depending on floor and usage conditions, the Scotch-Brite™ Diamond Floor Brush may last 18 times longer than competitive diamond floor pads.
Depending on many factors, including but not limited to: floor type, floor condition, pad pressure, water/chemical flow, chemical selection, and more, the Scotch-Brite™ Diamond Floor Brush can clean 280000 to 555000 sq m before needing to be replaced.
Standard sizes: Circular discs are available in a variety of common sizes. Brushes are approximately 25,4 mm thick
Storage: Store indoors in original packaging under normal climate controlled facility conditions
The Scotch-Brite™ Diamond Floor Brush is designed for use on uncoated* concrete floors. It is also suitable for use on uncoated* granite, epoxy based terrazzo, and cement based terrazzo floor surfaces. *Uncoated refers to floors without crystallization or topical coatings such as acrylic, urethane, or epoxy based coatings. Floors with a densifier or impregnator are okay for use with this system. The Scotch-Brite™ Diamond Floor Brush is used for routine cleaning and polishing under an auto scrubber or other standard low speed (175-600 RPM) floor cleaning equipment using water or a cleaning solution.
Applications:
• On low to medium gloss uncoated concrete floors that have not been polished recently: Use the Scotch-Brite™ diamond floor brush 1000 for daily cleaning. This is ideal for long-lasting cleaning and may provide gloss recovery.
• On uncoated concrete floors that have previously been polished with 3M™ Trizact™ Diamond TZ Abrasive Pads or similar methods: Use Scotch-Brite™ diamond floor brush 3000 for daily cleaning. This is ideal for long-lasting cleaning and for maintaining a high gloss with a possible gloss increase.
• To restore low gloss uncoated concrete floors to mediumhigh gloss: Begin with 3M™ Trizact™ diamond TZ Abrasive Pads, then switch to the Scotch-Brite™ diamond floor brush 3000. Ideal for restoring floor and providing long-lasting cleaning and maintaining high gloss with possible increase in gloss/DOI.
Verpakking: Each carton is labeled to identify the contents by product name, size, quantity, and manufacturer. Each brush does have a foam color identifier.