CRS Advanced Filter System Gas Purifiers
Leverancier: Trajan Scientific and Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
202903 202904 202905 202906 202912 202913 202914 202950 202955 202900MN 202902MN
SGEA202903EA 980 EUR
SGEA202903 SGEA202904 SGEA202905 SGEA202906 SGEA202912 SGEA202913 SGEA202914 SGEA202950 SGEA202955 SGEA202900MN SGEA202902MN
CRS Advanced Filter System Gas Purifiers
Filters Gasfilters
A high-capacity gas purifying system that can remove a combination of oxygen, moisture and hydrocarbons, (greater than C4) from inert gases and hydrogen.
  • Clean flow path (gas contacts only metal, glass and FKM seals)
  • High capacity and efficiency in a single cartridge
  • Easy replacement with twist on/off assembly knob
  • Double seal construction for safety
  • Check valves protect gas lines during replacement
  • Hardware for wall or bench mount
  • Stainless steel fittings
The AFS purifier I removes oxygen, moisture and hydrocarbons, (greater than C4) from inert gases and hydrogen. It contains high-sensitivity oxygen and moisture indicator.When cartridge is depleted the oxygen indicator changes from green to grey. The Moisture indicator changes from blue to brown. Both AFS purifier I and AFS purifier II have a very high capacity and can be expected to supply four GC's for at least one year when used with 99.995% pure gas supplies (provided reasonable care is taken during tank changes).
The advanced filter system complete kit includes the AFS cartridge, manifold, and the wall and bench mounting brackets.
A high-capacity, non-hazardous, gas purifying system designed to reduce contaminants in gases commonly used in laboratory applications to low-ppb levels, available in two models.