Vespel® GC Ferrules
Leverancier: Trajan Scientific and Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
212100 212103 212104 212105 212108 212110 212112 212124 212125 212164 212165 212168 2121XX 212200 212201 212205 212208 212210 212222 212228 2122XX 212400 212406 212410 212420 2124XX 212204
SGEA212100EA 53.3 EUR
SGEA212100 SGEA212103 SGEA212104 SGEA212105 SGEA212108 SGEA212110 SGEA212112 SGEA212124 SGEA212125 SGEA212164 SGEA212165 SGEA212168 SGEA2121XX SGEA212200 SGEA212201 SGEA212205 SGEA212208 SGEA212210 SGEA212222 SGEA212228 SGEA2122XX SGEA212400 SGEA212406 SGEA212410 SGEA212420 SGEA2124XX SGEA212204
Vespel® GC Ferrules
Fittingen en connectoren
Vespel GC ferrules are made from 100% high-temperature polyimide, a hard polymeric material that tends to seal permanently to capillary columns when in use. A firm torque is required to compress and seal.
  • Made from high temperature Dupont polyimide
  • They are not reusable
  • Vespel tends to shrink through repeated heating and cooling cycles
  • Hardest of the polymeric ferrules so a firm torque is required to compress and seal
They are excellent for creating seals on metal or glass and have long lifetimes. 100% Vespel is essentially non-porous to oxygen so it is ideal for GC/MS interface use, where air permeation can raise the background signal and contribute to phase degradation. They also work best in connections that are isothermal or insulated from oven temperature changes.
These ferrules are rated for use up to 350 °C.