CRS ZPure™ H₂O Indicating Filters
Leverancier: Trajan Scientific and Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
202234-B 202234-B-QC 202234RP-B-QC 202234RP-SS-QC 202234-SS 202234-SS-QC 202235-B 202235-B-QC 202235-SS 202235-SS-QC
SGEA202234-BEA 178 EUR
SGEA202234-B SGEA202234-B-QC SGEA202234RP-B-QC SGEA202234RP-SS-QC SGEA202234-SS SGEA202234-SS-QC SGEA202235-B SGEA202235-B-QC SGEA202235-SS SGEA202235-SS-QC
CRS ZPure™ H₂O Indicating Filters
Filters Gasfilters
The ZPure™ H₂O filter removes moisture to low ppb levels. The Co-free moisture indicator signals breakthrough when it changes from green to blue.
  • Capacity, H₂O: 6 g
  • Efficiency, H₂O: <20 ppb