CRS SPure™ H₂O/HC Filters
Leverancier: Trajan Scientific and Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
202324-4-PC 202324-6-PC 202324-B 202324-B-QC 202324D-4-PC 202324D-6-PC 202324D-B 202324D-B-QC 202324DRP-B-QC 202324DRP-SS-Q 202324D-SS 202324D-SS-QC 202324L-4-PC 202324L-6-PC 202324L-B 202324L-B-QC 202324LRP-B-QC 202324LRP-SS-Q 202324L-SS 202324L-SS-QC 202324RP-B-QC 202324RP-SS-QC 202324-SS 202324-SS-QC 202324XL-4-PC 202324XL-6-PC 202324XL-B 202324XL-B-QC 202324XLRP-B-Q 202324XLRP-SS- 202324XL-SS 202324XL-SS-QC 202325-B 202325-B-QC 202325D-B 202325D-B-QC 202325D-SS 202325D-SS-QC 202325L-B 202325L-B-QC 202325L-SS 202325L-SS-QC 202325-SS 202325-SS-QC 202325XL-B 202325XL-B-QC 202325XL-SS 202325XL-SS-QC
SGEA202324-4-PCEA 140 EUR
SGEA202324-4-PC SGEA202324-6-PC SGEA202324-B SGEA202324-B-QC SGEA202324D-4-PC SGEA202324D-6-PC SGEA202324D-B SGEA202324D-B-QC SGEA202324DRP-B-QC SGEA202324DRP-SS-Q SGEA202324D-SS SGEA202324D-SS-QC SGEA202324L-4-PC SGEA202324L-6-PC SGEA202324L-B SGEA202324L-B-QC SGEA202324LRP-B-QC SGEA202324LRP-SS-Q SGEA202324L-SS SGEA202324L-SS-QC SGEA202324RP-B-QC SGEA202324RP-SS-QC SGEA202324-SS SGEA202324-SS-QC SGEA202324XL-4-PC SGEA202324XL-6-PC SGEA202324XL-B SGEA202324XL-B-QC SGEA202324XLRP-B-Q SGEA202324XLRP-SS- SGEA202324XL-SS SGEA202324XL-SS-QC SGEA202325-B SGEA202325-B-QC SGEA202325D-B SGEA202325D-B-QC SGEA202325D-SS SGEA202325D-SS-QC SGEA202325L-B SGEA202325L-B-QC SGEA202325L-SS SGEA202325L-SS-QC SGEA202325-SS SGEA202325-SS-QC SGEA202325XL-B SGEA202325XL-B-QC SGEA202325XL-SS SGEA202325XL-SS-QC
CRS SPure™ H₂O/HC Filters
Filters Gasfilters
The SPure H₂O/HC Filter removes moisture, oil, dust, and hydrocarbons (C5 and heavier) from dry air and inert gases.
The cobalt-free moisture indicator signals breakthrough at 5% relative humidity and changes from green to blue.
SPure Filters have the option of standard compression fittings and quick connect fittings in ¹/₄ and ¹/₈" brass and stainless steel, or metal push-to-connect fittings in ¹/₄" and 6 mm for plastic tubing and non-carrier gas lines.