General Purpose Blue GC Septas
Leverancier: Trajan Scientific and Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
234106 234113 234108 234111 234116 234118 234121 234126 234216 234626 234906 234105 234109 234110 234115 234117 234120 234125 234215 234625 234905
SGEA234106EA 59.1 EUR
SGEA234106 SGEA234113 SGEA234108 SGEA234111 SGEA234116 SGEA234118 SGEA234121 SGEA234126 SGEA234216 SGEA234626 SGEA234906 SGEA234105 SGEA234109 SGEA234110 SGEA234115 SGEA234117 SGEA234120 SGEA234125 SGEA234215 SGEA234625 SGEA234905
General Purpose Blue GC Septas
Tussenschoten
General purpose blue GC septa, maded of silicone combines significantly longer injection life, low bleed and low injection port adhesion.
  • Reliabily seal against the carrier gas pressure in the inlet
  • Capable of being pierced and resealing time after time without pieces of the septa being deposited in the GC inlet system
  • Does not contaminate or bleed material into the chromatographic system
Has good durability, resealing, tear resistance and excellent solvent resistance.
Maximum injector temperature is 275 °C.
Verpakking: Packaged in glass jars for purity.