CRS 300 HC Filters
Leverancier: Trajan Scientific and Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
202330-B 202330-SS 202332-B 202332-SS
SGEA202330-BEA 173 EUR
SGEA202330-B SGEA202330-SS SGEA202332-B SGEA202332-SS
CRS 300 HC Filters
Filters Gasfilters
  • Capacity: 45 g
  • Efficiency: <5 ppb