Urine Bottles, Square
Leverancier: KARTELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0047900 0048000
KART0047900EA 225 EUR
KART0047900 KART0048000
Urine Bottles, Square
Diagnostische systemen Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
Bottle for clearance testing on urine collected during the whole day long.
  • Rectangular base for easy storage and enhanced stability in use
  • Ergonomic handgrip allows easy pouring
Adapted to contain a human biological sample (urine) in order to perform diagnostic laboratory analysis.
Certificaten: CE marked.