Dosipet Dispensers
Leverancier: KARTELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8307000 8307200 8308000 8308100
KART8307000EA 717 EUR
KART8307000 KART8307200 KART8308000 KART8308100
Dosipet Dispensers
Flesdispensers
Suitable for measuring out organic polar and non- polar solvents, including halogenated solvents and concentrated mineral acids, (including particularly volatile acids such as nitric and trifluoroacetic acids).
  • Can be used with peroxides, hydrochloric acid, and sulphuric acid
  • Borosilicate glass piston
  • Reagents come into contact only with parts made of PTFE
Available in multiple versions, depending on the reagents to be dispensed. The Dosipet S is useful for dosing concentrated acids, polar and non-polar solvents. The Dosipet HF is specially developed for hydrofluoric acid dosing. They are supplied complete with tubes for filling/discharging, as well as with a set of adaptors.