96 Well Plate, RCH TRACE-N
Leverancier: Tecan
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TCAT610-0101RCHEA 0 EUR
TCAT610-0101RCH
96 Well Plate, RCH TRACE-N
Microplaten Platen voor vastefase-extractie
End-capped C18 chemistry.
  • Reversed-phase
  • Liquid handler compatible
A 10 µm micro particulate with end-capped C18 chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample prep.