COURIER™ 3000 Shipping Scales
Leverancier: OHAUS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
30746021 30746022 30746023 30746025 30809911 30809912 30809913 30809914 30809915
OHAU30746021EA 240 EUR
OHAU30746021 OHAU30746022 OHAU30746023 OHAU30746025 OHAU30809911 OHAU30809912 OHAU30809913 OHAU30809914 OHAU30809915
COURIER™ 3000 Shipping Scales
Balansen en Weegschalen
The Courier 3000 Series shipping scales intuitive, user-friendly interfaces, bright displays, and low-profile steel construction ensure durability and reliability from offices to warehouses to even veterinary offices (the Courier 3000 is effective for weighing small animals). Versatile and effective.
  • For Weighing and display hold
  • 4-digit, 7-segment LCD display with white backlight, 28 mm high digits
  • ABS indicator, powder-coated steel platform and frame
  • 150% overload protection
  • AC adapter included
  • 100 Hours with disposable batteries
  • Painted steel platform construction