Ready-to-Load™ kits, amplification of DNA for fingerprinting
Leverancier: EDVOTEK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
130 130-C
EDVO130EA 109 EUR
EDVO130 EDVO130-C
Ready-to-Load™ kits, amplification of DNA for fingerprinting
Educatief materiaal Educatief materiaal over forensische wetenschappen Forensische kits
Ready-to-Load™ kit for teaching DNA fingerprinting and to show how DNA evidence is used in modern forensics. This experiment allows for varied results depending on the selection of DNA fingerprinting patterns.
  • For 6 lab groups
  • Complete in 50 minutes
Also required: Electrophoresis tank, power supply, automatic micropipette with tips, balance, microwave or hot plate, 65 °C water bath, white light visualisation system.
Leveringsinformatie: Kit includes instructions, Ready-to-Load™ DNA samples, agarose powder, practice gel loading solution, electrophoresis buffer, InstaStain® Methylene Blue, Methylene Blue Plus liquid stain, and microtipped transfer pipettes.