Ready-to-Load™ kits, DNA paternity test
Leverancier: EDVOTEK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
114 114-C
EDVO114EA 109 EUR
EDVO114 EDVO114-C
Ready-to-Load™ kits, DNA paternity test
Educatief materiaal Educatief materiaal over biologie Leeractiviteiten voor genetica
This experiment introduces students to the use of DNA fingerprinting in a simulated paternity determination. A child's DNA fingerprint is compared with his parents.
  • For 6 lab groups
Also required: Electrophoresis tank, power supply, micropipettes (5 to 50 µl adjustable or 40 µl fixed volume) with tips, and a white light box is recommended.
Leveringsinformatie: Kit includes instructions, Ready-to-Load™ DNA samples, agarose powder, practice gel loading solution, electrophoresis buffer, InstaStain® DNA stain, 1 ml calibrated drop pipette, 100 ml graduated cylinder and microtipped transfer pipettes.