Test tubes with side tube
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
92236900100 92237200100 92237300100
HECH92236900100EA 83.4 EUR
HECH92236900100 HECH92237200100 HECH92237300100
Test tubes with side tube
Buisjes Reageerbuisjes
DURAN® borosilicate glass, round bottom, beaded rim.
Test tubes with side tube (35×8 mm Ø) positioned 20 mm below rim.
Order Now
Specificaties
Environmentally Preferable