Analytical Membrane Bands
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
DS0600-0102 DS0600-0108 DS0600-0145 DS0600-0202 DS0600-0208 DS0600-0245
514-1430EA 455 EUR
514-1430 514-1431 514-1432 514-1433 514-1434 514-1435
Analytical Membrane Bands
Filters Membraanfilters
Analytical membrane bands are perfect for both microbial quality assurance (QA) and quality control (QC) applications where membrane dispensers are used.
  • Membranes made of MCE material
  • Bands are numbered to indicate how many membranes remain to be dispensed
  • Membranes are gridded to improve accuracy and save time
Compatible with most common dispensers on the market, including MilliporeSigma™ and Sartorius™, giving added flexibility and security of supply. Boxes feature two marked orientations and tear-away perforations with a thumb-tab for quick and easy loading.
Certificaten: Product has been validated to the highest standards, including ISO 7704 and ASTM F838.
Verpakking: Each case of pre-sterilised analytical membrane bands contains four boxes of 150 membranes per pack (600/case total).