Reusable Front Fitting Full Face Masks, FF-600F
Leverancier: 3M
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7100265074 7100265075 7100265236
MMMA7100265074EA 341 EUR
MMMA7100265074 MMMA7100265075 MMMA7100265236
Reusable Front Fitting Full Face Masks, FF-600F
Ademhalingsmaskers Herbruikbare maskers Volgelaatsmaskers
The FF-600F series reusable full face masks feature a panoramic visor that allows for a virtually unrestricted field of vision. Compatible with the 3M™ DT series filters, these face masks are suitable for a wide range of wearers and applications.
  • Panoramic visor provides a virtually unrestricted field of vision
  • Visor is designed to minimise visual distortion
  • Soft, highly flexible, and conforming Liquid Silicone Rubber faceseal enhances user comfort
  • Broad faceseal, designed to minimise pressure points and provide an effective seal without over-tightening the head harness
  • Matte finish TPE inner mask helps to prevent glare on the inside of the visor
  • Inner mask directional airflow helps prevent visor misting
  • Adjustable 5-point head harness enables a more tailored fit
  • Front-fitting filter port
  • Previously named the Scott™ Vision 2
These reusable full face masks feature a panoramic visor that is designed to prevent visual distortion and allows for a virtually unrestricted field of vision. The front-fitting filter port is compatible with 3M™ DT series filters. Please note, the FF-600F series reusable full face masks were previously named the Scott™ vision 2 reusable full face mask.