Chemical Spoon and Spatulas
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A625-150 A625-280 A625-320
SAMPA625-150EA 7.7 EUR
SAMPA625-150 SAMPA625-280 SAMPA625-320
Chemical Spoon and Spatulas
Lepels
Versatile spoon and spatula for sampling powder and liquid products.
  • Available in a range of sizes
  • Heavy duty design for comfortable use
  • Can be used as both a spoon and a spatula
  • Perfect for measuring, sampling and dosing powders and liquids
This versatile product has a robust design and is made from 304 stainless steel. It can be used as both a spoon and a spatula allowing greater versatility in the sampling of both powder and liquid products. There are no sharp edges or crevices and are ideal for use in food, pharma and chemical sectors.