Stainless Steel Tweezers
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A394-100 A394-200 A394-300 A394-400
SAMPA394-100EA 4.2 EUR
SAMPA394-100 SAMPA394-200 SAMPA394-300 SAMPA394-400
Stainless Steel Tweezers
Pincetten
These stainless steel tweezers are perfect for use in laboratory and production environments.
  • Made from 304 stainless steel
  • Available in a range of different lengths
  • Robust design
  • Useful for handling intricate items such as tissues
This range of stainless steel tweezers are perfect for a variety of applications across a wide range of industries including pharmaceutical. They are great for handling small items such as tissues and vials and are made from medical grade stainless steel.