SteriWare Swabbing Templates
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8888H-100 8888H-100S 8888H-50 8888H-50S
SAMP8888H-100EA 356 EUR
SAMP8888H-100 SAMP8888H-100S SAMP8888H-50 SAMP8888H-50S
SteriWare Swabbing Templates
Monsternemers Monsterdompelaars
Swabbing templates ensure a consistent swabbing area for cleaning validation purposes.
  • Choice of sterile or non sterile
  • Ensure consistent validation results
  • Individually bagged to reduce contamination
These swabbing templates are made in a cleanroom and come individually bagged to help reduce the risk of contamination.