Hand Scrapers
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8336P-1 8336R-1 8336B-1
SAMP8336P-1EA 31.3 EUR
SAMP8336P-1 SAMP8336R-1 SAMP8336B-1
Hand Scrapers
Schrapers
These robust hand scrapers are perfect for scraping large areas.
  • Made from high quality PP
  • Robust and incorporates a large sturdy handle
  • Large 250 mm wide blade
  • Comes in three distinctive colours
The range of robust and sturdy hand scrapers from Sampling Systems are perfect for easily scraping large areas due to their large blade size and robust handle. The performance is further enhanced by the fact it is made out of PP giving it very good chemical resistance.