Labels for BBP33 Printers, B-717
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
153151 153154 153153 153155 153159 153150 153156
BRDY153151EA 972.32 EUR
BRDY153151 BRDY153154 BRDY153153 BRDY153155 BRDY153159 BRDY153150 BRDY153156
Labels for BBP33 Printers, B-717
Etiketten
Static dissipative polyimide (B-717) withstands wave solder environments and multiple harsh washes for printed circuit board and electronic component preprocess labelling.
  • Service temperature of –70 to 100 °C
High temperature, glossy white polyimide material features a static dissipative adhesive and static dissipative release liner. Surface resistivity values in the recommended range for Dissipative ESD packaging materials as defined by ANSI/ESD S541-2008
Certificaten: UL Approved; meets requirements of MIL-STD-202G, method 215K, Compliant with Directive 2011/65/EU and 2015/863