uvex silv-Air classic Disposable Particle Filtering Respirators, FFP3
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8762310 8762312
UVEX8762310EA 79.5 EUR
UVEX8762310 UVEX8762312
uvex silv-Air classic Disposable Particle Filtering Respirators, FFP3
Ademhalingsmaskers Wegwerpmaskers Stofmaskers
uvex silv-Air particle filtering preformed mask with exhalation valve with level of protection FFP3.
  • Protection against harmful substances up to 30 times the limit value
  • Soft material edges for better wearer comfort
  • 4-point headband attachment for comfortable, individual and secure fit of the mask
  • Exhalation valve to reduce breathing resistance and to reduce heat and moisture under the mask
  • Integrated, flexible and adjustable nose clip
  • Compatible with uvex safety eyewear
  • All-around sealing lip for high levels of wearer comfort and a reliably secure fit
The respiratory protection mask can be used against solid, aqueous and nonvolatile liquid aerosols up to 30 times the
respective limit values. Mask fulfils the requirements for high dust exposure (D-label, dolomite dust test).
Area of use: Woodworking, hard wood, chromium, asbestos (short periods of work), stainless steel, lead, welding, dust that contain beryllium, arsenic, cadmium or cobalt. Medical waste disposal, fungal spores, legionella bacteria, infections, viruses, bacteria, enzyme, nanoparticles.
Certificaten: EN 149:2001+A1:2009.