HexArmor Helix® 3033 Cut Protection Gloves
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6066806 6066807 6066808 6066809 6066810 6066811
UVEX6066806EA 9.3 EUR
UVEX6066806 UVEX6066807 UVEX6066808 UVEX6066809 UVEX6066810 UVEX6066811
HexArmor Helix® 3033 Cut Protection Gloves
Handschoenen
HexArmor Helix 3033 gloves are 13-gauge gloves made of HPPE and steel fibers with 360° cut protection.
  • Seamless glove for increased comfort and breathability
  • Ultimate mobility and tactility
  • Suitable for dry and slightly damp areas
  • Provide abrasion resistance
  • Pre-shrunk and pre-washed
An elastic knit cuff prevents the penetration of dirt in the glove. Can be used directly contact with food or as underglove with more appropriate coating solution be used. These protective gloves are tight knitted and consist of strong and extremely abrasion-resistant fibres.
These protective gloves are tight knitted and consist of strong and extremely abrasion-resistant fibres. These comfortable knit gloves are a seamless series used for most tasks.
Certificaten: Outer layer tested according to FDA 21 CFR 177.1630 for food contact.