Hydrometers for Mineral Oil
Leverancier: Amarell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H838142-FL H838148-FL H838140-FL H838144-FL H838146-FL H838150-FL H838380-FL H838382-FL H838384-FL H838386-FL H838388-FL H838390-FL H838392-FL H838146FL/DKD3 H838148FL/DKD3
AMARH838142-FLEA 93.6 EUR
AMARH838142-FL AMARH838148-FL AMARH838140-FL AMARH838144-FL AMARH838146-FL AMARH838150-FL AMARH838380-FL AMARH838382-FL AMARH838384-FL AMARH838386-FL AMARH838388-FL AMARH838390-FL AMARH838392-FL AMARH838146FL/DKD3 AMARH838148FL/DKD3
Hydrometers for Mineral Oil
Hydrometers
High-precision hydrometers for the density range of 0,610 g/cm³ up to 1,100 g/cm³ (=g/ml); all instruments are permitted for government verification or government calibration; in general available from stock; further quality certificates on request.
  • Hydrometers with thermometer
  • Paper scale with well visible print
  • Shot weighted body
  • Accuracy ±1 scale division
  • Hermometer inside the hydrometer body
  • Capillary prismatic
  • Filling: Paraffine oil (Pet)
  • Capillary filling back: clear
Verpakking: Packed in a transparent plastic case.