TEGERA® 295 Waterproof Winter Gloves
Leverancier: EJENDALS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
295-10 295-12 295-7 295-8
EJEN295-10EA 153.42 EUR
EJEN295-10 EJEN295-12 EJEN295-7 EJEN295-8
TEGERA® 295 Waterproof Winter Gloves
Handschoenen
TEGERA® 295 gloves are professional waterproof winter glove for cold and wet outdoor work.
  • Durable
  • Good fingertip sensitivity
  • Extra flexible, perfect fit with good grip
  • Extra comfortable
  • Warm lining and windproof
  • Reinforced fingers and thumb
  • Type of work: Medium weight
The ergonomic design provides a superior all-day comfort, reducing hand fatigue. The combination of durable leather and 40 g Thinsulate™ technology makes it the ideal glove for cold insulation in many different robust jobs.
These gloves can be used while kachine operating, building and construction, carpentry, transport work, machine driving, airport work, repair work and prevents the risk of contact with dirt, abrasion injuries, scratches, lacerations, grazes, contact with cold, contact with damp, contact with moisture, blisters.
Colour: White, grey, blue
Material details:
Palm: Full grain goatskin
Back: Spandex
Inner: Polyester
Middle: TPU (Thermoplastic polyurethane)
Outer: Leather, elastane, polyester, natural latex
Certificaten: EN ISO 21420:2020, EN 388:2016+A1:2018 2121X, Cat. II , EN 511:2006 121