DuraClone Dry Unitized Antibody Panels For Flow Cytometry
Leverancier: Beckman Coulter
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B53309 B53318 B53328 B53346 C00162 C34369 C49589 C97741
BECMB53309EA 1290 EUR
BECMB53309 BECMB53318 BECMB53328 BECMB53346 BECMC00162 BECMC34369 BECMC49589 BECMC97741
DuraClone Dry Unitized Antibody Panels For Flow Cytometry
Testen Cellulaire assays
The dry DURAClone antibody panels are designed for clinical research studies. Every assay tube or well contains a layer with the dried-down antibody panel unitized for a single test.
  • Dry format, unitized for a single test, no antibody or stimulant pipetting
  • Flexible by optional addition of liquid reagents
  • Color compensation solution included (where required)
  • Expert-proven and thoroughly tested
  • Storage&shipping at room temperature
  • Simplified inventory management
Antibody panel development requires a high level of flow cytometry expertise and may consume critical laboratory resources. Take a leap forward and streamline panel implementation in your flow cytometry assay menu by using expert proven, dry DURAClone antibody combinations. The elimination of liquid antibody handling also efficiently reduces human error sources and reagent stability issues - the ideal solution for longitudinal and multi-center clinical research studies!
Certificaten: Although not required for RUO status, this product is manufactured in accordance with Good Manufacturing Practices.
Leveringsinformatie: Supplied with screw cap and pouring ring.
Verpakking: The bottles are packaged in shrinked-wrapped modules.