96- and 384-well cell culture microplates, Advanced TC™
Leverancier: Greiner Bio-One
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
655980_100 655982_160 655983_32 655986_32 675986_32 781983_32 781986_32 788986_60
392-0227EA 569 EUR
392-0227 392-0228 392-0229 392-0230 392-0248 392-0264 392-0265 392-0269
96- and 384-well cell culture microplates, Advanced TC™
Microplaten Celcultuurplaten
Microplates with a modified culture surface to optimise cell characteristics and functions. The improved adherence and increased growth rate improve and accelerate cell proliferation. Additionally, the Advanced TC™ surface facilitates uniform and consistent cell adherence, increases throughput and reduces cell loss, for example during automated washing stages. Free of detectable DNase, RNase and human DNA.
  • Increased cell adherence
  • Morphology similar to in vivo conditions
  • Optimal cultivation conditions for sensitive cells
  • Enables reduced quantities of serum or serum-free use
  • Free of cytotoxic substances