Experimentation kits, mitochondrial DNA analysis using PCR
Leverancier: EDVOTEK EUROPE LTD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EDVO332EA 287 EUR
EDVO332
Experimentation kits, mitochondrial DNA analysis using PCR
Educatief materiaal Educatief materiaal over biologie PCR-leeractiviteiten
The mitochondria are thought to have evolved from a symbiotic relationship between prokaryotic and eukaryotic cells. Mitochondria have their own DNA and are only inherited via the maternal line. In this experiment, students amplify two regions of their mitochondrial DNA.
  • For 25 students
  • Set up 30 minutes; PCR 2 hours or overnight; electrophoresis 45 minutes
Also required: Micropipettes (5 to 50 µl) with tips, water bath, thermal cycler, electrophoresis tank and power supply.
Leveringsinformatie: Kit includes instructions, proteinase K, PCR beads, control DNA and primers, microtubes, chelating agent, agarose, DNA ladder, practice gel loading solution, gel loading dye, electrophoresis buffer, and gel stain.