Ready-to-Load™ kits, what is PCR and how does it work?
Leverancier: Edvotek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Ready-to-Load™
EDVOS-48EA 93 EUR
EDVOS-48
Ready-to-Load™ kits, what is PCR and how does it work?
Educatief materiaal Educatief materiaal over biologie PCR-leeractiviteiten
This simulation experiment demonstrates the process of DNA amplification by PCR and how the amplified product is detected by separating the reaction mixture by agarose gel electrophoresis.
  • For 10 lab groups
  • Complete in 45 minutes
Also required: Electrophoresis tank and power supply.
Leveringsinformatie: Kit includes instructions, Ready-to-Load™ dye samples, agarose powder, practice gel loading solution, electrophoresis buffer, and microtipped transfer pipettes.