Experimentation kits, introduction to ELISA reactions
Leverancier: EDVOTEK EUROPE LTD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EDVO269EA 190 EUR
EDVO269
Experimentation kits, introduction to ELISA reactions
Educatief materiaal Educatief materiaal over biologie Physiology and Health Learning Activities
Also required: Distilled or deionised water, 37 °C incubator, automatic micropipettes with tips, and laboratory glassware.
Leveringsinformatie: Kit includes instructions, antigens, primary and secondary antibodies, peroxide co-substrate, hydrogen peroxide, ABTS substrate, phosphate buffered saline, tubes, plates, and transfer pipettes.