Experimentation kits, simulation of HIV detection by ELISA
Leverancier: EDVOTEK EUROPE LTD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EDVO271EA 190 EUR
EDVO271
Experimentation kits, simulation of HIV detection by ELISA
Educatief materiaal Educatief materiaal over biologie Physiology and Health Learning Activities
Also required: 37 °C incubator
Leveringsinformatie: Kit includes instructions, simulated HIV antigens, positive and negative controls, simulated donor serum, secondary antibody, substrate, microtitre plates, transfer pipettes, and microtubes.