Experimentation kits, principles of thin layer chromatography
Leverancier: Edvotek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EDVO113EA 169 EUR
EDVO113
Experimentation kits, principles of thin layer chromatography
Educatief materiaal Educatief materiaal over chemie Leeractiviteiten voor chromatografie
This experiment introduces chromatographic theory and methods of thin layer chromatography (TLC). A mixture of dyes are separated on a cellulose-based TLC plate using different solvent systems.
  • For 8 lab groups
  • Spotting plates 20 minutes, TLC separation 5 minutes
Also required: 5ml or 10 ml pipettes.
Leveringsinformatie: Kit includes instructions, samples, reagents and solvents, cellulose TLC plate, 5 µl capillary pipettes.