Experimentation kits, ion exchange chromatography
Leverancier: Edvotek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EDVO243EA 181 EUR
EDVO243
Experimentation kits, ion exchange chromatography
Educatief materiaal Educatief materiaal over chemie Leeractiviteiten voor chromatografie
Most molecules have a net charge within a pH range of 2 to 10. When the pH is altered, the net charge on molecules can change drastically. In this experiment, a mixture of two chemicals is absorbed onto a solid support ion-exchange column and separated during elution under conditions that influence their net charge.
  • For 6 lab groups
  • Requires 60 to 90 minutes
Also required: Spectrophotometer and cuvettes, ring stands and clamps, and 5 ml pipettes.
Leveringsinformatie: Kit includes instructions, ion exchanger, chemical mixture, potassium acetate buffer, and chromatography columns.