Capillary tube on foot
Leverancier: Hecht
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
201-0216EA 46.4 EUR
201-0216
Capillary tube on foot
Educatief materiaal Educatief materiaal over fysica Leeractiviteiten voor mechanica
Five communicating glass tubes with different inner diameters. Used to show capillary forces and how they are affected by the inner diameter of the pipes.
Leveringsinformatie: Comes with foot.