Safety Harnesses, XE200
Leverancier: 3M
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7100316788 7100316790
MMMA7100316788EA 378 EUR
MMMA7100316788 MMMA7100316790
Safety Harnesses, XE200
Valbescherming Valharnassen
3M DBI-SALA ExoFit XE200 harness with rescue loops, automatic buckles, two sizes. X-shaped chest straps with central D-ring attachment point optimised position on the body. Revolver adjuster for the torso length for a quick, secure and comfortable fit of the harness.
  • Auto-locking quick connect
  • Length: 260 mm
  • Fall arrest loop placement: Back, front, shoulder
  • Product colour: grey
  • Integrated flat suspension trauma step loops
  • Reusable and repositionable lanyard holders for quick and easy parking of carabiners
The XE200 comfort harness consists of a durable and hard-wearing polyester webbing, lightweight aluminium mounting components, robust (yet lightweight) polyester hybrid padding with additional air channels and reflective 3M Scotchlite inserts. It also has a turret adjuster for the torso length and self-locking automatic buckles to optimise the fit. The 3M DBI-SALA ExoFit XE200 is compatible with a range of 3M fall protection accessories for tools. All models are equipped with suspension trauma step loops.
Self-locking automatic buckle ensures secure contact with the harness webbing. An integrated pHSG tunnel enables quick and easy connection and disconnection of personal fall arresters.