Flame Resistant Anti-Static, High Visibility Fleece Jackets
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FR31YERL FR31YERM FR31YERS FR31YERXL FR31YERXXL FR31YERXXXL
PRTWFR31YERLEA 145 EUR
PRTWFR31YERL PRTWFR31YERM PRTWFR31YERS PRTWFR31YERXL PRTWFR31YERXXL PRTWFR31YERXXXL
Flame Resistant Anti-Static, High Visibility Fleece Jackets
Werkkleding
This inherently flame resistant two-tone fleece offers exceptional warmth and comfort. The high quality anti-pill finish maintains the look of the garment for longer.
  • Front zip opening for easy access
  • Contrast fleece collar for protection against dirt
  • Contrast colouring for added style
  • 40+ UPF rated fabric to block 98% of UV rays
  • Premium sew on flame resistant reflective tape
  • Two pockets for secure storage
The features include contrast panels for protection against dirt, two side zipped pockets, elasticated cuffs and an adjustable drawcord hem.
Shell fabric : Modaflame fleece HV: 60% modacrylic, 39% cotton, 1% carbon fibre 280 g
Certificaten: EN ISO 11612 A1, B1, C2, EN 1149 -5, IEC 61482-2 IEC 61482-1-1 ELim 5.9CAL/CM², IEC 61482-2 IEC 61482-1-2 APC 1, EN ISO 20471 Class 3
Opgelet: Do not bleach, dry clean, or tumble dry.