Versilon™ PTFE Tubings
Leverancier: Saint Gobain Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A210059010010A A210048008010A A210030010010B A210040010010B A200010005010A A210025007510B
VERNA210059010010AEA 532 EUR
VERNA210059010010A VERNA210048008010A VERNA210030010010B VERNA210040010010B VERNA200010005010A VERNA210025007510B
Versilon™ PTFE Tubings
Slangen Transfer Tubing
Versilon™ PTFE Flexible Tubing from Saint-Gobain has the highest working temperature and chemical resistance of any fluoropolymer tubing available today.
  • Fully fluorinated fluoropolymer tubing
  • Opaque/translucent colour, other colours available on request
  • Highest working temperature (up to 260 °C) out of all fluoropolymer tubing
  • Autoclavable
This tubing is ideal for use in general chemical transfer applications such as delivery of natural gas and mineral oils, toxic gas monitoring, and paint, varnish, or adhesive delivery lines. It also serves numerousMore Product Information
Order Now
Specificaties
Model VERNA210059010010A VERNA210048008010A VERNA210030010010B VERNA210040010010B VERNA200010005010A VERNA210025007510B
Environmentally Preferable
Maximumtemperatuur 327 °C 327 °C 260 °C 260 °C 260 °C 327 °C
Min. temperatuur −200 °C −200 °C −200 °C −200 °C −200 °C −200 °C