Permanent Autoclave-Resistant Thermal Transfer Labels
Leverancier: GA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AUTC-258C1-3BL AUTC-258C1-3GA AUTC-258C1-3RE AUTC-258C3-6BL AUTC-258C3-6GA AUTC-258C3-6RE AUTC75NPC1-2GA AUTC75NPC1-2OR AUTC75NPC3-2GA AUTC75NPC3-2OR AUTC84C10.25BL AUTC84C10.25GA AUTC84C10.25OR AUTC84C10.25PI AUTC84C10.25RE AUTC84C10.25YE AUTC-84C3-1BL AUTC-84C3-1GA AUTC-84C3-1OR AUTC-84C3-1PI AUTC-84C3-1RE AUTC-84C3-1YE AUTT-103C1-2WH AUTT-103C3-2WH AUTT-105C1-2WH AUTT-105C3-2WH AUTT-19C1-1WH AUTT-19C3-1WH AUTT-200C1-1WH AUTT-200C3-2WH AUTT216C10.5WH AUTT-216C3-1WH AUTT-231C3-2WH AUTT-258C1-3WH AUTT-258C3-6WH AUTT-27C1-1WH AUTT-27C3-2WH AUTT-28C1-2WH AUTT-28C3-2WH AUTT-36C1-1WH AUTT62C1-0.5WH AUTT-62C3-1WH AUTT75NPC1-2WH AUTT75NPC3-2WH AUTT84C10.25WH AUTT-84C3-1WH
GAINAUTC-258C1-3BLEA 168 EUR
GAINAUTC-258C1-3BL GAINAUTC-258C1-3GA GAINAUTC-258C1-3RE GAINAUTC-258C3-6BL GAINAUTC-258C3-6GA GAINAUTC-258C3-6RE GAINAUTC75NPC1-2GA GAINAUTC75NPC1-2OR GAINAUTC75NPC3-2GA GAINAUTC75NPC3-2OR GAINAUTC84C10.25BL GAINAUTC84C10.25GA GAINAUTC84C10.25OR GAINAUTC84C10.25PI GAINAUTC84C10.25RE GAINAUTC84C10.25YE GAINAUTC-84C3-1BL GAINAUTC-84C3-1GA GAINAUTC-84C3-1OR GAINAUTC-84C3-1PI GAINAUTC-84C3-1RE GAINAUTC-84C3-1YE GAINAUTT-103C1-2WH GAINAUTT-103C3-2WH GAINAUTT-105C1-2WH GAINAUTT-105C3-2WH GAINAUTT-19C1-1WH GAINAUTT-19C3-1WH GAINAUTT-200C1-1WH GAINAUTT-200C3-2WH GAINAUTT216C10.5WH GAINAUTT-216C3-1WH GAINAUTT-231C3-2WH GAINAUTT-258C1-3WH GAINAUTT-258C3-6WH GAINAUTT-27C1-1WH GAINAUTT-27C3-2WH GAINAUTT-28C1-2WH GAINAUTT-28C3-2WH GAINAUTT-36C1-1WH GAINAUTT62C1-0.5WH GAINAUTT-62C3-1WH GAINAUTT75NPC1-2WH GAINAUTT75NPC3-2WH GAINAUTT84C10.25WH GAINAUTT-84C3-1WH
Permanent Autoclave-Resistant Thermal Transfer Labels
Etiketten
Permanent autoclave-resistant labels withstand various sterilisation procedures, including in steam autoclaves, dry heat ovens, gamma irradiators, and ethylene oxide gas sterilisers.
  • Temperature range: −60 to 150 °C
  • Ideal for use in repeated steam autoclave sterilization cycles, gamma irradiation (up to 50 KGy), and ethylene oxide sterilisation
  • Printable with all major thermal transfer printer brands
  • Waterproof and weatherproof
  • RR-class resin ribbon recommended
High temperature-resistant permanent labels for identifying various sterilisation containers and surfaces that will undergo autoclaving and dry heat sterilisation. These labels can be applied to metal, plastic, and glass surfaces, and will withstand up to 20 autoclave sterilisation cycles at 121 °C and 16,5 psi. The labels and the printout can withstand high temperature exposure, as well as gamma irradiation (up to 50 KGy) and ethylene oxide sterilisation. Ideal for identifying containers and surfaces that need to be sterilised inside autoclaves, dry heat sterilising equipment, and with gamma irradiators.