Adflo™ Powered Air Purifying Respirator Systems with Speedglas™ G5-01 Series Welding Helmet
Leverancier: 3M
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7100256131 7100257943
MMMA7100256131EA 2580 EUR
MMMA7100256131 MMMA7100257943
Adflo™ Powered Air Purifying Respirator Systems with Speedglas™ G5-01 Series Welding Helmet
Ademhalingsmaskers Ademhalingssystemen
The Adflo™ Powered air purifying respirator system with 3M™ Speedglas™ G5-01 series Welding helmet provides all of the benefits of the G5-01 series welding helmet with a convenient Adflo™ Powered air purifying respirator system.
  • Robust, compact design
  • Ergonomic, adjustable head suspension
  • 73×109 mm viewing area
  • Flip-front welding shield allows for a clear, wide-view protective visor for grinding and setup
  • Available in two different auto-darkening lens options (VC/TW)
  • Audible and visual alarms signal low airflow and low battery charge
  • G5 series storage/carry bag included
For workers in heavy-duty environments, the Adflo™ Powered air purifying respirator with Speedglas™ G5-01 series Welding helmet system provides high-efficiency particulate respiratory protection for welders in a robust yet compact design with ergonomic, adjustable head suspension. Flip-front welding shield allows for a clear, wide-view protective visor for grinding and setup. The Adflo™ Powered air respirator has a high-efficiency particulate filter that can be 'stacked' onto an optional Adflo™ filter (not included) for additional protection against certain vapours or a Nuisance odour pad assembly. Audible and visual alarms signal low airflow and low battery charge.
Recommended applications include metal repair, metal processing, grinding, and welding.