DURAN® Glass Capillaries
Leverancier: Schott AG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1026406 1026409 1026413 1026415 1026419 1026405 1026408 1026410 1026685 1026399 1026690 1026417 1026411 1026404 1026412 1026414 1026418 1026420 1026421 1026422 1026398 1026400 1026401 1026397 1026396 1026402 1027407 1024403 1036416
SCOR1026406EA 1570 EUR
SCOR1026406 SCOR1026409 SCOR1026413 SCOR1026415 SCOR1026419 SCOR1026405 SCOR1026408 SCOR1026410 SCOR1026685 SCOR1026399 SCOR1026690 SCOR1026417 SCOR1026411 SCOR1026404 SCOR1026412 SCOR1026414 SCOR1026418 SCOR1026420 SCOR1026421 SCOR1026422 SCOR1026398 SCOR1026400 SCOR1026401 SCOR1026397 SCOR1026396 SCOR1026402 SCOR1027407 SCOR1024403 SCOR1036416
DURAN® Glass Capillaries
Pipetten Micro-capillairen
High precision capillaries made from borosilicate 3.3 glass.
  • High chemical resistance
  • High hydrolytical resistance
  • High temerature resistance
  • High resistance to thermal shock