DYMO Compatible Cryogenic Labels for Frozen Surfaces
Leverancier: GA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EDCS-081WH EDCS-040WH EDCS-070WH EDCS-082WH
GAINEDCS-081WHEA 79.8 EUR
GAINEDCS-081WH GAINEDCS-040WH GAINEDCS-070WH GAINEDCS-082WH
DYMO Compatible Cryogenic Labels for Frozen Surfaces
Etiketten
DYMO compatible labels for already frozen surfaces. These labels will adhere to frozen vials and containers under deep-freeze conditions (−80 °C).
  • Quick labelling of frozen vials and tubes (no thawing required)
  • Withstands exposure to solvents, water baths, and condensation
  • Compatible with the 450 series and 4XL DYMO® LabelWriters
  • Convenient to print; no need for ribbons or other ink source
LabTAG’s unique and innovative CryoSTUCK® technology is now available for labels compatible with DYMO 450 LabelWriter printers. These solvent-resistant cryogenic labels are specially designed to be directly applied to already frozen surfaces like vials and tubes stored under deep freeze conditions (−80 °C). These labels help preserve the integrity of valuable temperature-sensitive samples and specimens, since there is no need to thaw the containers before attaching them to the frozen vial. They come with a permanent adhesive that will keep the label from falling off or flagging over time under liquid nitrogen’s conditions. Once the labels are applied, the containers/vials can be returned for long-term preservation in liquid nitrogen (−196 °C) or ultra-low temperature freezers (−80 °C). These labels are ideal for usage in research institutes, clinical laboratories, biobanks, biotechnology companies, and more.
In addition to their CryoSTUCK® features, these DYMO compatible labels display excellent resistance to routinely used laboratory chemicals like detergents, alcohols (100% Ethanol immersion: up to 15 minutes), acetone, xylene (immersion: up to 30 minutes), disinfecting solutions, as well as surface-sanitising wipes.